Learn More
Invitrogen™ GALNT13 Monoclonal Antibody (3F3)
Mouse Monoclonal Antibody
Supplier:  Invitrogen™ MA522929
Catalog No. PIMA522929
Only null left
Add to Cart
Description
Description
Peptide Sequence: RAVISRNQEG PGEMGKAVLI PKDDQEKMKE LFKINQFNLM ASDLIALNRS LPDVRLEGCK TKVYPDELPN TSVVIVFHNE AWSTLLRTVY SVINRSPHYL LSEVI.
The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GALNT13
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.4
Q8IUC8
GALNT13
GALNT13 (NP_443149.1, 47 a.a. ~ 151 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
100 μg
Primary
Human
Antibody
IgG2b κ
ELISA, Western Blot
3F3
Unconjugated
GALNT13
A230002A12; A230020F20; BB182356; FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; Galnt13; H_NH0187G20.1; KIAA1918; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; polypeptide N-acetylgalactosaminyltransferase 13; pp-GalNAc-T13; pp-GaNTase 13; protein-UDP acetylgalactosaminyltransferase 13; T13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1
Mouse
Affinity chromatography
RUO
114805
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.