Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA543522
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA543522 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA543522 Supplier Invitrogen™ Supplier No. PA543522
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: CNKCDDKKER SLLPALRAVI SRNQEGPGEM GKAVLIPKDD QEKMKELFKI Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.The GALNT13 protein is a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAcT; EC 2.4.1.41) family, which initiate O-linked glycosylation of mucins (see MUC3A, MIM 158371) by the initial transfer of N-acetylgalactosamine (GalNAc) with an alpha-linkage to a serine or threonine residue.[supplied by OMIM].
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT13
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene GALNT13
Gene Accession No. Q8IUC8
Gene Alias A230002A12; A230020F20; BB182356; FLJ16031; FLJ41157; GalNAc transferase 13; GalNAc-T13; Galnt13; H_NH0187G20.1; KIAA1918; MGC119459; MGC119461; Polypeptide GalNAc transferase 13; polypeptide N-acetylgalactosaminyltransferase 13; pp-GalNAc-T13; pp-GaNTase 13; protein-UDP acetylgalactosaminyltransferase 13; T13; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 13 (GalNAc-T13); WUGSC:H_NH0187G20.1
Gene Symbols GALNT13
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human GALNT13 (aa 31-80).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114805
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.