Learn More
Invitrogen™ GALNT14 Monoclonal Antibody (2A8)

Description
Peptide Sequence: KNGDDRQQWT KTGSHIEHIA SHLCLDTDMF GDGTENGKEI VVNPCESSLM SQHWDMVS.
Specifications
Specifications
| Antigen | GALNT14 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 2A8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | Galnt14 |
| Gene Accession No. | Q96FL9 |
| Gene Alias | 0610033M06Rik; FLJ12691; FLJ13977; GalNac-T10; GalNac-T14; GALNT14; GALNT15; Polypeptide GalNAc transferase 14; polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; protein-UDP acetylgalactosaminyltransferase 14; si:ch211-147c1.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14); UNQ2434/PRO4994 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.