Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT14 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113171
Description
GALNT14 Polyclonal Antibody for Western Blot, IHC (P), ELISA
GALNT14 (EC 2.4.1.41) belongs to a large subfamily of glycosyltransferases residing in the Golgi apparatus. GALNT enzymes catalyze the first step in the O-glycosylation of mammalian proteins by transferring N-acetyl-D-galactosamine (GalNAc) to peptide substrates.
Specifications
| GALNT14 | |
| Polyclonal | |
| Unconjugated | |
| Galnt14 | |
| 0610033M06Rik; FLJ12691; FLJ13977; GalNac-T10; GalNac-T14; GALNT14; GALNT15; Polypeptide GalNAc transferase 14; polypeptide N-acetylgalactosaminyltransferase 14; polypeptide N-acetylgalactosaminyltransferase 15; pp-GaNTase 14; protein-UDP acetylgalactosaminyltransferase 14; si:ch211-147c1.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 14; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase T10; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 14 (GalNAc-T14); UNQ2434/PRO4994 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 313878, 71685, 79623 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8BVG5, Q96FL9 | |
| Galnt14 | |
| Recombinant Human Polypeptide N-acetylgalactosaminyltransferase 14 protein (1-552AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction