Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT16 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563666
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563666 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563666 Supplier Invitrogen™ Supplier No. PA563666
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATSTLMSSPG SPVILQMCNP REGKQKWRRK GSFIQHSVSG LCLETKPAQL VTSKCQADAQ AQQWQLLPH Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 94%.

GALNTL1 belongs to the glycosyltransferase 2 family, GalNAc-T subfamily. It contains 1 ricin B-type lectin domain. GALNTL1 may catalyze the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT16
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT16
Gene Accession No. Q8N428
Gene Alias 5730405L21Rik; AI415388; GALNACT16; galNAc-T16; GalNAc-T-like protein 1; GALNT16; GALNTL1; KIAA1130; MGC141855; mpp-GalNAc-T16; Polypeptide GalNAc transferase 16; polypeptide GalNAc transferase-like protein 1; Polypeptide N-acetylgalactosaminyltransferase 16; polypeptide N-acetylgalactosaminyltransferase-like 1; polypeptide N-acetylgalactosaminyltransferase-like protein 1; pp-GaNTase-like protein 1; protein-UDP acetylgalactosaminyltransferase-like protein 1; putative polypeptide N-acetylgalactosaminyltransferase-like protein 1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 1; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 16; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 1
Gene Symbols GALNT16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT16. Recombinant protein control fragment (Product #RP-101145).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57452
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.