Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121044
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121044 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121044 Supplier Invitrogen™ Supplier No. PA5121044
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, Jurkat, HepG2, mouse lung. The target is usually found in the following locations: Golgi apparatus, Golgi stack membrane, Secreted, Single-pass type II membrane protein. Immunogen sequence: HQDIAFGALQ QGTNCLDTLG HFADGVVGVY ECHNAGGNQE WALTKEKSVK HMDLCLTVVD RAPGSLIKLQ GCRENDSRQK WEQIEGNSKL RHVGSNLCLD SRTAKSGGLS VEVCGPALSQ QWKFTLNLQQ.

Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Has a broad spectrum of substrates for peptides such as EA2, Muc5AC, Muc1a, Muc1b. Probably involved in O-linked glycosylation of the immunoglobulin A1 (IgA1) hinge region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.08 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GALNT2
Gene Accession No. Q10471, Q6PB93
Gene Alias AI480629; E4TF1B; GABP subunit beta-1; GABP subunit beta-2; GABPB; GABPB1; GABPB-1; GABPB2; GABPB-2; GalNAc-T2; galnt2; im:7140357; polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; ppGaNTase-T2; protein-UDP acetylgalactosaminyltransferase 2; RP5-956O18.1; si:dkey-162b3.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2)
Gene Symbols GALNT2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 442-571 of human GALNT2 (NP_004472.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108148, 2590, 292090
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.