Learn More
Invitrogen™ GALNT2 Recombinant Rabbit Monoclonal Antibody (011)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA530452
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human GALNT2/GalNAc-T2.
Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. Has a broad spectrum of substrates for peptides such as EA2, Muc5AC, Muc1a, Muc1b. Probably involved in O-linked glycosylation of the immunoglobulin A1 (IgA1) hinge region.
Specifications
GALNT2 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q10471 | |
GALNT2 | |
Recombinant Human GALNT2/GalNAc-T2 Protein (Lys52-Gln571). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
ELISA | |
011 | |
Unconjugated | |
GALNT2 | |
AI480629; E4TF1B; GABP subunit beta-1; GABP subunit beta-2; GABPB; GABPB1; GABPB-1; GABPB2; GABPB-2; GalNAc-T2; galnt2; im:7140357; polypeptide GalNAc transferase 2; Polypeptide N-acetylgalactosaminyltransferase 2; Polypeptide N-acetylgalactosaminyltransferase 2 soluble form; pp-GaNTase 2; ppGaNTase-T2; protein-UDP acetylgalactosaminyltransferase 2; RP5-956O18.1; si:dkey-162b3.1; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2 (GalNAc-T2) | |
Rabbit | |
Protein A | |
RUO | |
2590 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.