Learn More
Invitrogen™ GALNT3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589560
Description
Positive Samples: HT-29, BT-474, MCF-7, SW480, Mouse kidney; Cellular Location: Golgi apparatus, Golgi stack membrane, Single-pass type II membrane protein Immunogen sequence: VQYSKEESRM ERNMKNKNKM LDLMLEAVNN IKDAMPKMQI GAPVRQNIDA GERPCLQGYY TAAELKPVLD RPPQDSNAPG ASGKAFKTTN LSVEEQKEKE RGEAKHCFNA FASDRISL.
GALNT3 encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein is highly homologous to other family members, however the enzymes have different substrate specificities.
Specifications
GALNT3 | |
Polyclonal | |
Unconjugated | |
Galnt3 | |
GalNAc transferase 3; galNAc-T3; GALNT3; HFTC; HHS; polypeptide GalNAc transferase 3; polypeptide GalNAc-transferase T3; polypeptide N-acetylgalactosaminyltransferase 3; Polypeptide N-acetylgalactosaminyltransferase 3-like protein; pp-GaNTase 3; ppGaNTase-T3; protein-UDP acetylgalactosaminyltransferase 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); polypeptide N-acetylgalactosaminyltransferase 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
14425, 2591, 366061 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.32 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P70419, Q14435 | |
Galnt3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 43-160 of human GALNT3 (NP_0044732). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.