Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALNT3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526217
Description
GALNT3 encodes UDP-GalNAc transferase 3, a member of the GalNAc-transferases family. This family transfers an N-acetyl galactosamine to the hydroxyl group of a serine or threonine residue in the first step of O-linked oligosaccharide biosynthesis. Individual GalNAc-transferases have distinct activities and initiation of O-glycosylation is regulated by a repertoire of GalNAc-transferases. The protein is highly homologous to other family members, however the enzymes have different substrate specificities.
Specifications
| GALNT3 | |
| Polyclonal | |
| Unconjugated | |
| Galnt3 | |
| GalNAc transferase 3; galNAc-T3; GALNT3; HFTC; HHS; polypeptide GalNAc transferase 3; polypeptide GalNAc-transferase T3; polypeptide N-acetylgalactosaminyltransferase 3; Polypeptide N-acetylgalactosaminyltransferase 3-like protein; pp-GaNTase 3; ppGaNTase-T3; protein-UDP acetylgalactosaminyltransferase 3; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 3 (GalNAc-T3); polypeptide N-acetylgalactosaminyltransferase 3 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 2591 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.37 mg/mL | |
| PBS with 0.09% sodium azide | |
| Q14435 | |
| Galnt3 | |
| KLH conjugated synthetic peptide between 454-481 amino acids from the central region of human GALNT3 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction