Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5100537
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5100537 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5100537 Supplier Invitrogen™ Supplier No. PA5100537
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GALNT4.

This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. In vitro, the encoded protein can complement other GalNAc-Ts in the complete O-glycosylation of the mucin-1 tandem repeat and can O-glycosylate the P-selectin glycoprotein ligand-1 molecule. The coding region of this gene is contained within a single exon.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GALNT4
Gene Accession No. O08832, Q8N4A0
Gene Alias AV011803; GALNACT4; GalNAc-T4; Galnt4; Polypeptide GalNAc transferase 4; polypeptide GalNAc transferase-T4; polypeptide N-acetylgalactosaminyltransferase 4; pp-GaNTase 4; ppGaNTase-T4; protein-UDP acetylgalactosaminyltransferase 4; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 4 (GalNAc-T4)
Gene Symbols GALNT4
Host Species Rabbit
Immunogen A synthesized peptide derived from human GALNT4(Accession Q8N4A0), corresponding to amino acid residues P438-Y488.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14426, 500826, 8693
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.