Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552641
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552641 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552641 Supplier Invitrogen™ Supplier No. PA552641
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MKPPLRGHGK GAWGKENVRK TEESVLKVEV DLDQTQRERK MQNALGRGKV VPLWHPAHLQ TLPVTPNKQK TDGRGTKPEA SSHQGTPKQT TAQGAPKTSF IAAKGTQVVK ISVHMGRVSL KQEPR Highest antigen sequence identity to the following orthologs: Mouse - 51%, Rat - 50%.

GALNT5 can catalyze the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor. GALNT5 has activity toward EA2 peptide substrate, but it has a weak activity toward Muc2 or Muc1b substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT5
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT5
Gene Accession No. Q7Z7M9
Gene Alias GALNACT5; GalNAc-T5; GALNT5; Polypeptide GalNAc transferase 5; polypeptide N-acetylgalactosaminyltransferase 5; pp-GaNTase 5; protein-UDP acetylgalactosaminyltransferase 5; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 5; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 5 (GalNAc-T5)
Gene Symbols GALNT5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT5. Recombinant protein control fragment (Product #RP-90245).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11227
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.