Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590082
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590082 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590082 Supplier Invitrogen™ Supplier No. PA590082
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DVSSREEATE KPWLKSLVSR KDHVLDLMLE AMNNLRDSMP KLQIRAPEAQ QTLFSINQSC LPGFYTPAEL KPFWERPPQD PNAPGADGKA FQKSKWTPLE TQEKEEGYKK H; Positive Samples: U-87MG, BxPC-3; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.

This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. The encoded protein is capable of glycosylating fibronectin peptide in vitro and is expressed in a fibroblast cell line, indicating that it may be involved in the synthesis of oncofetal fibronectin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT6
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.93 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GALNT6
Gene Accession No. Q8NCL4
Gene Alias 4632410F13; AW047994; fc23d02; fc56f12; GalNAc transferase 6; GalNAcT6; galNAc-T6; GALNT6; N-acetylgalactosaminyltransferase T-6; polypeptide GalNAc transferase 6; Polypeptide N-acetylgalactosaminyltransferase 6; pp-GaNTase; pp-GaNTase 6; protein-UDP acetylgalactosaminyltransferase 6; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6); wu:fc23d02; wu:fc56f12; zgc:77836
Gene Symbols GALNT6
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human GALNT6 (NP_0091412).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11226
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.