Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GALNT6 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA528064
Description
Recommended positive controls: 293T, A431, H1299, Molt-4, PC-12. Predicted reactivity: Mouse (92%), Rat (91%), Xenopus laevis (80%), Chicken (82%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase (GalNAc-T) family of enzymes. GalNAc-Ts initiate mucin-type O-linked glycosylation in the Golgi apparatus by catalyzing the transfer of GalNAc to serine and threonine residues on target proteins. They are characterized by an N-terminal transmembrane domain, a stem region, a lumenal catalytic domain containing a GT1 motif and Gal/GalNAc transferase motif, and a C-terminal ricin/lectin-like domain. GalNAc-Ts have different, but overlapping, substrate specificities and patterns of expression. The encoded protein is capable of glycosylating fibronectin peptide in vitro and is expressed in a fibroblast cell line, indicating that it may be involved in the synthesis of oncofetal fibronectin.
Specifications
GALNT6 | |
Polyclonal | |
Unconjugated | |
GALNT6 | |
4632410F13; AW047994; fc23d02; fc56f12; GalNAc transferase 6; GalNAcT6; galNAc-T6; GALNT6; N-acetylgalactosaminyltransferase T-6; polypeptide GalNAc transferase 6; Polypeptide N-acetylgalactosaminyltransferase 6; pp-GaNTase; pp-GaNTase 6; protein-UDP acetylgalactosaminyltransferase 6; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 6 (GalNAc-T6); wu:fc23d02; wu:fc56f12; zgc:77836 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100361647, 11226 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.79 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q8NCL4 | |
GALNT6 | |
Recombinant fragment corresponding to a region within amino acids 30 and 518 of Human GALNT6. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction