Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA564213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA564213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA564213 Supplier Invitrogen™ Supplier No. PA564213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DDPSPLSRMR EDRDVNDPMP NRGGNGLAPG EDRFKPVVPW PHVEGVEVDL ESIRRINKAK NEQEHHAGG Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 93%.

This gene encodes GalNAc transferase 7, a member of the GalNAc-transferase family. The enzyme encoded by this gene controls the initiation step of mucin-type O-linked protein glycosylation and transfer of N-acetylgalactosamine to serine and threonine amino acid residues. This enzyme is a type II transmembrane protein and shares common sequence motifs with other family members. Unlike other family members, this enzyme shows exclusive specificity for partially GalNAc-glycosylated acceptor substrates and shows no activity with non-glycosylated peptides. This protein may function as a follow-up enzyme in the initiation step of O-glycosylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT7
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT7
Gene Accession No. Q86SF2
Gene Alias AI225872; GalNAcT7; GalNAc-T7; Galnt7; N-acetylgalactosaminyltransferase 7; polypeptide GalNAc transferase 7; polypeptide N-acetylgalactosaminyltransferase 7; pp-GaNTase 7; ppGaNTase-T7; protein-UDP acetylgalactosaminyltransferase 7; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 7; UDP-N-acetyl-alpha-D-galactosamine: polypeptide N-acetylgalactosaminyltransferase 7; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 7 (GalNAc-T7)
Gene Symbols GALNT7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNT7. Recombinant protein control fragment (Product #RP-103595).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51809
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.