Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNT8 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567291
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567291 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567291 Supplier Invitrogen™ Supplier No. PA567291
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: QMKLFPHSQLF RQWGEDLSEA QQKAAQDLFR KFGYNAYLSN QLPLNRTIPD TRDYRCLRKT YPSQLPSLSV I Highest antigen sequence identity to the following orthologs - mouse 40%, rat 40%.

Probably catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNT8
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT8
Gene Accession No. Q9NY28
Gene Alias GalNAc transferase 8; GalNAc-T8; GALNT8; Polypeptide GalNAc transferase 8; polypeptide N-acetylgalactosaminyltransferase 8; pp-GaNTase 8; probable polypeptide N-acetylgalactosaminyltransferase 8; Protein-UDP acetylgalactosaminyltransferase 8; UDP-GalNAc: polypeptide N-acetylgalactosaminyltransferase 8; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 8; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 8 (GalNAc-T8)
Gene Symbols GALNT8
Host Species Rabbit
Immunogen Recombinant Human GALNT8. Recombinant protein control fragment (Product #RP-107930).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26290
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.