Learn More
Invitrogen™ GALNTL4 Monoclonal Antibody (1F9)

Description
Peptide Sequence: SQQIHVGILS PTVDDDDNRC LVDVNSRPRL TECSYAKAKR MKLHWQFSQG GPIQNRKSKR CLELQENSDL EFGFQLVLQK CSGQHWSITN VLRSL*.
Specifications
Specifications
| Antigen | GALNTL4 |
| Applications | ELISA |
| Classification | Monoclonal |
| Clone | 1F9 |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | GALNT18 |
| Gene Accession No. | Q6P9A2 |
| Gene Alias | 2900011G21Rik; BC024988; GalNAc-T15; GALNACT18; GalNAc-T18; GalNAc-T-like protein 4; GalNAc-transferase 18; GALNT15; GALNT18; Galntl4; LOW QUALITY PROTEIN: polypeptide N-acetylgalactosaminyltransferase 18; mpp-GalNAc-T18(O18); polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; polypeptide N-acetylgalactosaminyltransferase 18; polypeptide N-acetylgalactosaminyltransferase-like 4; polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; RGD1309623; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4 |
| Gene Symbols | GALNT18 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.