Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNTL4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553017 Supplier Invitrogen™ Supplier No. PA553017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SDIIAYGVLQ NSLKTDLCLD QGPDTENVPI MYICHGMTPQ NVYYTSSQQI HVGILSPTVD DDDNRCLVDV NSRPRLIECS YAKAKRMKLH WQFSQGGPIQ NRKSKRCLEL QENSDLEFGF QLVLQKCSGQ HWSITNVL Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 95%.

Catalyzes the initial reaction in O-linked oligosaccharide biosynthesis, the transfer of an N-acetyl-D-galactosamine residue to a serine or threonine residue on the protein receptor.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNTL4
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GALNT18
Gene Accession No. Q6P9A2
Gene Alias 2900011G21Rik; BC024988; GalNAc-T15; GALNACT18; GalNAc-T18; GalNAc-T-like protein 4; GalNAc-transferase 18; GALNT15; GALNT18; Galntl4; LOW QUALITY PROTEIN: polypeptide N-acetylgalactosaminyltransferase 18; mpp-GalNAc-T18(O18); polypeptide GalNAc transferase 18; Polypeptide GalNAc transferase-like protein 4; polypeptide N-acetylgalactosaminyltransferase 18; polypeptide N-acetylgalactosaminyltransferase-like 4; polypeptide N-acetylgalactosaminyltransferase-like protein 4; pp-GaNTase-like protein 4; protein-UDP acetylgalactosaminyltransferase-like protein 4; putative polypeptide N-acetylgalactosaminyltransferase-like protein 4; RGD1309623; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase-like protein 4; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 4
Gene Symbols GALNT18
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALNTL4. Recombinant protein control fragment (Product #RP-90092).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 374378
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.