Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALNTL5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116326
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116326 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116326 Supplier Invitrogen™ Supplier No. PA5116326
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

GALNTL5 Polyclonal Antibody for Western Blot

Probable inactive glycosyltransferase required during spermatid development. May participate in protein loading into the acrosomes and accumulation of ubiquitin-proteasome systems around the head-tail coupling apparatus region.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALNTL5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene GALNTL5
Gene Accession No. Q7Z4T8
Gene Alias galNAc-T15; GALNACT19; GalNAc-T5L; GALNT15; GALNTL5; Inactive polypeptide N-acetylgalactosaminyltransferase-like protein 5; polypeptide GalNAc transferase 15; polypeptide GalNAc transferase-like 5; polypeptide N-acetylgalactosaminyltransferase-like 5; pp-GaNTase 15; protein-UDP acetylgalactosaminyltransferase 15; putative polypeptide N-acetylgalactosaminyltransferase-like protein 5; testis tissue sperm-binding protein Li 61n; UDP-GalNAc:polypeptide N-acetylgalactosaminyltransferase 15; UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase-like 5
Gene Symbols GALNTL5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 52-80 amino acids from the N-terminal region of human GALNTL5.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168391
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.