Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALR1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533570
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533570 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533570 Supplier Invitrogen™ Supplier No. PA533570
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Percent identity with other species by BLAST analysis: Gorilla (100%) Human, Chimpanzee, Orangutan, Gibbon (95%) Monkey (89%).

The neuropeptide galanin elicits a range of biological effects by interaction with specific G-protein-coupled receptors. Galanin receptors are seven-transmembrane proteins shown to activate a variety of intracellular second-messenger pathways. GALR1 inhibits adenylyl cyclase via a G protein of the Gi/Go family. GALR1 is widely expressed in the brain and spinal cord, as well as in peripheral sites such as the small intestine and heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALR1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene GALR1
Gene Accession No. P47211
Gene Alias Gal receptor 1; Gal-1 receptor; GAL1-R; Galanin r1 receptor; galanin receptor 1; galanin receptor type 1; Galanin-1 receptor; Galn1r; GALNR; Galnr1; Galr1; Gal-r1; GALR-1; Hugal 1-r
Gene Symbols GALR1
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from C-terminus of human GALR1.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.