Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALR2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533574
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533574 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533574 Supplier Thermo Scientific Supplier No. PA533574
Add to Cart
Add to Cart

GALR2 Polyclonal Antibody

GALR2 Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen GALR2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Galr2
Gene Accession No. O43603
Gene Alias Gal receptor 2; Gal-2 receptor; GAL2-R; Galanin r2 receptor; galanin receptor 2; galanin receptor type 2; Galanin-2 receptor; Galnr2; GALR2; Gal-r2; GALR-2; mGalR; MGC125983; MGC125984
Gene Symbols Galr2
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from C-terminus of human GALR2.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8811
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.