Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALR2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA577441
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Per vendor datasheet, reconstitute with 50uL deionized water.
Galanin is an important neuromodulator present in the brain, gastrointestinal system, and hypothalamopituitary axis. It is a 30-amino acid non-C-terminally amidated peptide that potently stimulates growth hormone secretion, inhibits cardiac vagal slowing of heart rate, abolishes sinus arrhythmia, and inhibits postprandial gastrointestinal motility. The actions of galanin are mediated through interaction with specific membrane receptors that are members of the 7-transmembrane family of G protein-coupled receptors. GALR2 interacts with the N-terminal residues of the galanin peptide. The primary signaling mechanism for GALR2 is through the phospholipase C/protein kinase C pathway (via Gq), in contrast to GALR1, which communicates its intracellular signal by inhibition of adenylyl cyclase through Gi. However, it has been demonstrated that GALR2 couples efficiently to both the Gq and Gi proteins to simultaneously activate 2 independent signal transduction pathways.
Specifications
GALR2 | |
Polyclonal | |
Unconjugated | |
GALR2 | |
GALR2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
14428, 29234, 8811 | |
-20°C | |
Lyophilized |
Immunofluorescence, Immunohistochemistry, Western Blot | |
0.75 mg/mL | |
PBS with 1% BSA and 0.05% sodium azide; pH 7.4 | |
O08726, O43603, O88854 | |
Galr2 | |
Peptide (C)GSGSQRAKRKVTR, corresponding to amino acid residues 223-235 of mouse Galanin receptor type 2 | |
50 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction