Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALR3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA528747
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA528747 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA528747 Supplier Invitrogen™ Supplier No. PA528747
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, H1299, HeLa, HepG2, Molt-4, Raji, mouse eye, mouse brain, rat brain. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The neuropeptide galanin modulates a variety of physiologic processes including cognition/memory, sensory/pain processing, hormone secretion, and feeding behavior. The human galanin receptors are G protein-coupled receptors that functionally couple to their intracellular effector through distinct signaling pathways. GALR3 is found in many tissues and may be expressed as 1.4-, 2.4-, and 5-kb transcripts. GALR3 has been reported in brain, testis, adrenal gland, and pancreas. An EST has been isolated from an ovarian thecoma fibroma library.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALR3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.72 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene GALR3
Gene Accession No. O60755, O88626, O88853
Gene Alias Gal receptor-3; Gal3 receptor; GAL3-R; Galanin r3 receptor; galanin receptor 3; galanin receptor type 3; galanin receptor, family member 3; Galnr3; Galr3; Gal-r3; GALR-3; OTTHUMP00000028820
Gene Symbols GALR3
Host Species Rabbit
Immunogen Synthetic peptide encompassing a sequence within the Extracellular domain of human GALR3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14429, 29235, 8484
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.