Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALT Antibody (SR1733), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | GALT |
---|---|
Clone | SR1733 |
Dilution | Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
Applications | Western Blot, Immunofluorescence |
Classification | Monoclonal |
Description
GALT Monoclonal antibody specifically detects GALT in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
GALT | |
Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.7.12, Gal-1-P uridylyltransferase, galactose-1-phosphate uridyl transferase, galactose-1-phosphate uridylyltransferase, UDP-glucose--hexose-1-phosphate uridylyltransferase | |
A synthesized peptide derived from human GALT (Uniprot #: P07902) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1733 | |
Western Blot, Immunofluorescence | |
Unconjugated | |
Rabbit | |
metabolism | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
2592 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title