Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GALT Antibody (SR1733), R&D Systems™
SDP

Catalog No. NB260461 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260461 100 μg
NB260463 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260461 Supplier R&D Systems Supplier No. NBP322582100UL

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

GALT Monoclonal specifically detects GALT in Human∣Mouse∣Rat samples. It is validated for Immunocytochemistry/Immunofluorescence∣Western Blot.

Specifications

Antigen GALT
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone SR1733
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias EC 2.7.7.12, Gal-1-P uridylyltransferase, galactose-1-phosphate uridyl transferase, galactose-1-phosphate uridylyltransferase, UDP-glucose--hexose-1-phosphate uridylyltransferase
Host Species Rabbit
Immunogen A synthesized peptide derived from human GALT (Uniprot #: P07902)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline metabolism
Primary or Secondary Primary
Gene ID (Entrez) 2592
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.