Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GALT Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552125
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552125 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552125 Supplier Invitrogen™ Supplier No. PA552125
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RANDHQHIRY NPLQDEWVLV SAHRMKRPWQ GQVEPQLLKT VPRHDPLNPL CPGAIRANGE VNPQYDSTFL FDNDFPALQP DAPSPGPSDH PLFQAKSARG VCKVMCFHPW SD Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 91%.

Galactose-1-phosphate uridyl transferase (GALT) catalyzes the second step of the Leloir pathway of galactose metabolism, namely the conversion of UDP-glucose + galactose-1-phosphate to glucose-1-phosphate + UDP-galactose. The absence of this enzyme results in classic galactosemia in humans and can be fatal in the newborn period if lactose is not removed from the diet. The pathophysiology of galactosemia has not been clearly defined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GALT
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Galt
Gene Accession No. P07902
Gene Alias AW553376; Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; galactose-1-phosphate uridylyltransferase; galactose-1-phosphate uridylyltransferase isoform 1; GALT; IL11RA; interleukin 11 receptor, alpha; UDP-glucose--hexose-1-phosphate uridylyltransferase
Gene Symbols Galt
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GALT. Recombinant protein control fragment (Product #RP-102415).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2592
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.