Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GALT Polyclonal Antibody
GALT Polyclonal Antibody
Supplier: Thermo Scientific PA579298
Description
GALT Polyclonal Antibody for Western Blot

Specifications
GALT | |
Polyclonal | |
Unconjugated | |
Galt | |
AW553376; Gal-1-P uridylyltransferase; galactose-1-phosphate uridyl transferase; galactose-1-phosphate uridylyltransferase; galactose-1-phosphate uridylyltransferase isoform 1; GALT; IL11RA; interleukin 11 receptor, alpha; UDP-glucose--hexose-1-phosphate uridylyltransferase | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
2592, 298003 | |
-20°C | |
Lyophilized |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
500 μg/mL | |
PBS with 4mg trehalose and no preservative | |
P07902, P43424 | |
Galt | |
E. coli-derived human GALT recombinant protein (Position: Q188-A379). | |
100 μg | |
Primary | |
Human, Rat, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction