Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Galunisertib
GREENER_CHOICESDP

Catalog No. 69565 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
69-565 5 mg
69-562-5 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 69-565 Supplier Tocris Bioscience™ Supplier No. 6956/5
Add to Cart
Add to Cart
This item is not returnable. View return policy

ALK4 and ALK5 (TGFβRI) inhibitor

Galunisertib is an ALK4 and ALK5 (TGFβRI) inhibitor (IC50 values are 77.7 and 172 nM, respectively). Also inhibits a range of other kinases with submicromolar IC50s including MINK, TGFβRII, ALK6 and ACVR2B. Inhibits TGFβ-induced SMAD phosphorylation in cancer cell lines in vitro and in vivo. Also reverses TGFβ-mediated suppression of T cell proliferation. Prevents TGFβ-mediated EMT and cell migration in vitro and exhibits antitumor efficacy in vivo. Used as component of growth media for urothelial organoids. Orally bioavailable.

Specifications

Chemical Name or Material 4-[5,6-Dihydro-2-(6-methyl-2-pyridinyl)-4 H-pyrrolo[1,2-b]pyrazol-3-yl]-6-quinolinecarboxamide
CAS 700874-72-2
Quantity 5 mg
Target TGF-beta Receptor Inhibitors
Molecular Formula C22H19N5O
Formula Weight 369.43
Product Type Inhibitor
Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.