Learn More
Invitrogen™ gamma Actin Polyclonal Antibody

Description
Recommended positive controls: NIH-3T3. Predicted reactivity: Mouse (100%), Rat (100%), Xenopus laevis (100%), Dog (100%), Chicken (99%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Specifications
Specifications
| Antigen | gamma Actin |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.26 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 |
| Gene | ACTG1 |
| Gene Accession No. | P63260, P63261 |
| Gene Alias | ACT; ACTG; Actg1; actin; actin gamma; actin gamma 1; actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma, cytoplasmic 1; Actl; AL023024; beta actin; beta-actin FE-3; BRWS2; cytoplasmic 2; cytoplasmic ACTG1; cytoplasmic type 5; cytoskeletal gamma-actin; cytoskeleton actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E51; epididymis luminal protein 176; gamma 1 propeptide; gamma actin; gamma-actin; HEL-176; type 5 |
| Gene Symbols | ACTG1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.