Learn More
gamma Actin Polyclonal Antibody, Invitrogen™
Description
Suggested positive control: Hela whole cell extract.
Specifications
Specifications
| Antigen | gamma Actin |
| Applications | Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Formulation | tris citrate, tris phosphate with 0.09% sodium azide |
| Gene | ACTG1 |
| Gene Accession No. | P63261, P63260 |
| Gene Alias | ACT; ACTG; Actg1; actin; actin gamma; actin gamma 1; actin, cytoplasmic 2; Actin, cytoplasmic 2, N-terminally processed; Actin, cytoplasmic type 5; actin, cytoplasmic, type 5; actin, gamma 1; actin, gamma 1 propeptide; actin, gamma, cytoplasmic 1; Actl; AL023024; beta actin; beta-actin FE-3; BRWS2; cytoplasmic 2; cytoplasmic ACTG1; cytoplasmic type 5; cytoskeletal gamma-actin; cytoskeleton actin; deafness, autosomal dominant 20; deafness, autosomal dominant 26; DFNA20; DFNA26; E51; epididymis luminal protein 176; gamma 1 propeptide; gamma actin; gamma-actin; HEL-176; type 5 |
| Gene Symbols | ACTG1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.