Learn More
gamma Adaptin/AP1G1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
| Antigen | gamma Adaptin/AP1G1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Liquid |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-8632
|
Bethyl Laboratories, Inc
A304771AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Orangutan
Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is composed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. Two transcript variants encoding different isoforms have been found for this gene.Specifications
| gamma Adaptin/AP1G1 | |
| Polyclonal | |
| Liquid | |
| RUO | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| adapter-related protein complex 1 subunit gamma-1, adaptor protein complex AP-1 subunit gamma-1, adaptor-related protein complex 1 gamma 1 subunit, adaptor-related protein complex 1 subunit gamma-1, adaptor-related protein complex 1, gamma 1 subunit, ADTG, CLAPG1, clathrin assembly protein complex 1 gamma large chain, clathrin assembly protein complex 1 gamma-1 large chain, clathrin-associated/assembly/adaptor protein, large, gamma 1, gamma adaptin, gamma1-adaptin, golgi adaptor HA1/AP1 adaptin gamma subunit, golgi adaptor HA1/AP1 adaptin subunit gamma-1, testicular tissue protein Li 21 | |
| Between 1 and 50 | |
| Primary | |
| 4° C | |
| AP1G1 |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| O43747, P22892 | |
| 11765, 164 | |
| IgG | |
| Antigen affinity chromatography | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.