Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Adaptin Monoclonal Antibody (100/3), Invitrogen™

Catalog No. PIMA125066
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA125066 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA125066 Supplier Thermo Scientific Supplier No. MA125066

Mouse Monoclonal Antibody

Recommended positive controls: bovine brain. MA1-25066 will not react with mouse or rat. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

gamma Adaptin Monoclonal antibody specifically detects gamma Adaptin in Bovine, Human, Non-human Primate samples. It is validated for Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Adaptin
Applications Immunocytochemistry, Immunomicroscopy, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 100/3
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation Ascites with 0.1% sodium azide
Gene AP1G1
Gene Alias AA409002; adapter-related protein complex 1 subunit gamma-1; adaptor protein complex AP-1 gamma 1 subunit; adaptor protein complex AP-1 subunit gamma-1; adaptor protein complex AP-1, gamma 1 subunit; adaptor related protein complex 1 gamma 1 subunit; adaptor related protein complex 1 subunit gamma 1; adaptor-related protein complex 1 gamma 1 subunit; adaptor-related protein complex 1 subunit gamma-1; adaptor-related protein complex 1, gamma 1 subunit; ADTG; AP-1 complex subunit gamma-1; Ap1g1; AU041323; AW551707; CLAPG1; clathrin assembly protein complex 1 gamma large chain; clathrin assembly protein complex 1 gamma-1 large chain; clathrin-associated/assembly/adaptor protein, large, gamma 1; D8Ertd374e; gamma adaptin; Gamma1-adaptin; gamma-adaptin; golgi adaptor HA1/AP1 adaptin gamma subunit; golgi adaptor HA1/AP1 adaptin subunit gamma-1; MGC18255; testicular tissue protein Li 21
Gene Symbols AP1G1
Host Species Mouse
Immunogen AP-1 adaptor from bovine brain
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 507677
Target Species Bovine, Human, Non-human Primate
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.