Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ gamma Adducin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA536235
Description
This antibody detects endogenous protein at a molecular weight of 82 kDa. Purity is >95% by SDS-PAGE.
Adducins are heteromeric proteins composed of different subunits referred to as adducin alpha, beta and gamma. The three subunits are encoded by distinct genes and belong to a family of membrane skeletal proteins involved in the assembly of spectrin-actin network in erythrocytes and at sites of cell-cell contact in epithelial tissues. While adducins alpha and gamma are ubiquitously expressed, the expression of adducin beta is restricted to brain and hematopoietic tissues. Adducin, originally purified from human erythrocytes, was found to be a heterodimer of adducins alpha and beta. Polymorphisms resulting in amino acid substitutions in these two subunits have been associated with the regulation of blood pressure in an animal model of hypertension. Heterodimers consisting of alpha and gamma subunits have also been described. Structurally, each subunit is comprised of two distinct domains. The amino-terminal region is protease resistant and globular in shape, while the carboxy-terminal region is protease sensitive. The latter contains multiple phosphorylation sites for protein kinase C, the binding site for calmodulin, and is required for association with spectrin and actin. Alternatively spliced adducin gamma transcripts encoding different isoforms have been described. The functions of the different isoforms are not known.
Specifications
gamma Adducin | |
Polyclonal | |
Unconjugated | |
ADD3 | |
ADD3; ADDG; Addl; adducin 3; adducin 3 (gamma); Adducin gamma; adducin-like protein 70; AI463285; CPSQ3; cytoskeletal protein; gamma-adducin; potein kinase C binding protein 35H; protein kinase C-binding protein 35H; R75380 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
120, 25230, 27360 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
Q62847, Q9QYB5, Q9UEY8 | |
ADD3 | |
Synthetic peptide corresponding to amino acids 420-471 of human Adducin gamma. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction