Learn More
Gamma-Adducin Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303716AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Adducins are a family of cytoskeleton proteins encoded by three genes ( alpha, beta and gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localized at the spectrin-actin junction of the membrane skeleton. The cortical actin cytoskeletal network is lost during apoptosis and adducins are central in the cortical actin network organization. Adducin alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin alpha and Adducin gamma is ubiquitous in contrast to the restricted expression of Adducin beta. Adducin beta is expressed at high levels in brain andhematopoietic tissues, such as bone marrow in humans, and spleen in mice.Specifications
Gamma-Adducin | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9UEY8 | |
ADD3 | |
Between 650 and 700 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
ADD3 | |
ADD3, ADDG, ADDL, Adducin 3 (gamma), Adducin 3, Adducin-like protein 70, Gamma-adducin, Adducin gamma | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
120 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.