Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gamma-Adducin Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501565862
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-5862 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-5862 Supplier Bethyl Laboratories, Inc Supplier No. A303716A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Adducins are a family of cytoskeleton proteins encoded by three genes ( alpha, beta and gamma). Adducin is a protein associated with the inner leaflet of the plasma membrane and is one of the proteins localized at the spectrin-actin junction of the membrane skeleton. The cortical actin cytoskeletal network is lost during apoptosis and adducins are central in the cortical actin network organization. Adducin alpha is a cytoskeletal protein involved with sodium-pump activity in the renal tubule and is associated with hypertension. The expression of Adducin alpha and Adducin gamma is ubiquitous in contrast to the restricted expression of Adducin beta. Adducin beta is expressed at high levels in brain andhematopoietic tissues, such as bone marrow in humans, and spleen in mice.

Specifications

Antigen Gamma-Adducin
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ADD3
Gene Accession No. Q9UEY8
Gene Alias ADD3, ADDG, ADDL, Adducin 3 (gamma), Adducin 3, Adducin-like protein 70, Gamma-adducin, Adducin gamma
Gene Symbols ADD3
Host Species Rabbit
Immunogen Between 650 and 700
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 120
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.