Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody (15F11), DyLight 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP254527G
Description
gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.Specifications
gamma Catenin | |
Monoclonal | |
DyLight 488 | |
50 mM sodium borate with 0.05% sodium azide | |
JUP | |
Recombinant, full length chicken gamma Catenin protein (Uniprot: P14923) | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Feline | |
IgG1 κ |
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
15F11 | |
Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
Mouse | |
Protein A or G purified | |
Cancer, Cytoskeleton Markers, Phospho Specific | |
3728.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction