Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin Antibody (15F11), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00 - $524.00

Specifications

Antigen gamma Catenin
Clone 15F11
Concentration 0.2 mg/mL
Dilution Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB25332520
SDP
View Documents
Novus Biologicals
NBP25332520UG
20 μg
Each for $268.00
Only null left
Add to Cart
 
NB253325100
SDP
View Documents
Novus Biologicals
NBP253325100UG
100 μg
Each for $524.00
Only null left
Add to Cart
 
NB253325200
SDP
View Documents
Novus Biologicals
NBP253325200UG
200 μg N/A N/A N/A
Description

Description

gamma Catenin Monoclonal specifically detects gamma Catenin in Human, Rat, Bovine, Canine, Chicken, Feline samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

gamma Catenin
0.2 mg/mL
Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence
Unconjugated
Mouse
Cancer, Cytoskeleton Markers, Phospho Specific
10mM PBS with 0.05% BSA with 0.05% Sodium Azide
3728
Recombinant, full length chicken Catenin gamma protein
Primary
Store at 4C.
15F11
Western Blot 0.5 - 1.0 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat, Bovine, Canine, Chicken, Feline
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB
JUP
IgG1 κ
Protein A or G purified
84 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.