Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP323197
Description
gamma Catenin Polyclonal antibody specifically detects gamma Catenin in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
gamma Catenin | |
Polyclonal | |
Western Blot 1:1000 | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
gamma Catenin synthetic peptide (coupled to KLH) corresponding to amino acid residues from the C-terminal region of human gamma Catenin. This peptide sequence is highly conserved in rat and mouse gamma Catenin. [UniProt# P14923] | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
3728 | |
Store at -20°C. | |
IgG |
Western Blot | |
Unconjugated | |
PBS, 1 mg/ml BSA and 50% glycerol | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction