Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody (rCTNG/1664), DyLight 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308469DL594
Description
gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
gamma Catenin | |
Monoclonal | |
DyLight 594 | |
catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
Recombinant human gamma Catenin protein (Uniprot: P14923) | |
100 μg | |
Cancer, Cell Biology, Cellular Markers | |
3728 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
rCTNG/1664 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction