Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Catenin Antibody (rCTNG/1664), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308469IR
Description
gamma Catenin Monoclonal antibody specifically detects gamma Catenin in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| gamma Catenin | |
| Monoclonal | |
| DyLight 755 | |
| catenin (cadherin-associated protein), gamma (80kD), catenin (cadherin-associated protein), gamma 80kDa, Catenin gamma, Desmoplakin III, Desmoplakin-3, DP3CTNNG, DPIII, gamma-catenin, junction plakoglobin, PDGBARVD12, PKGB | |
| Recombinant human gamma Catenin protein (Uniprot: P14923) | |
| 100 μg | |
| Cancer, Cell Biology, Cellular Markers | |
| 3728 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| rCTNG/1664 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction