Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

gamma Catenin (JUP) Monoclonal Antibody (OTI4D5), TrueMAB™, OriGene
SDP

Catalog No. 502543678
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-3678 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-3678 Supplier OriGene Supplier No. CF812398
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

gamma Catenin (JUP) Monoclonal antibody specifically detects gamma Catenin (JUP) in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen gamma Catenin (JUP)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4D5
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene JUP
Gene Accession No. P14923, Q02257, Q6P0K8
Gene Alias ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 500-745 of human JUP (NP_002221) produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16480, 3728, 81679
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.