Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Catenin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595204
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA595204 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA595204 Supplier Invitrogen™ Supplier No. PA595204
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Human gamma Catenin shares 98% amino acid (aa) sequence identity with both mouse and rat gamma Catenin. Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human 293T whole cell, human MCF-7 whole cell, human T47D whole cell, rat heart tissue. IHC: human intestinal cancer tissue, mouse intestine tissue, rat intestine tissue IHC-F: rat intestine tissue. ICC/IF: A431 cell, human MCF-7 cell. Flow: A431 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Catenin
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 5mg BSA and 0.05mg sodium azide
Gene JUP
Gene Accession No. P14923, Q02257, Q6P0K8
Gene Alias ARVD12; catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; cb80; cell-adhesion protein plakoglobin; Ctnng; D930025P04Rik; danPG; Desmoplakin III; desmoplakin-3; desmosomal protein; DP3; DPIII; fb11b06; gama-catenin; gamma-catenin; gamma-catenin (plakoglobin); junction plakoglobin; junction plakoglobin a; Jup; jup protein; jupa; OTTHUMP00000164735; PDGB; PG; PKGB; plakoglobin; wu:fb11b06
Gene Symbols JUP
Host Species Rabbit
Immunogen E.coli-derived human gamma Catenin recombinant protein (Position: M556-A745).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16480, 3728, 81679
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.