Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SCNN1G Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110347 Supplier Thermo Scientific Supplier No. PA5110347
Add to Cart
Add to Cart

SCNN1G Polyclonal Antibody

SCNN1G Polyclonal Antibody for Western Blot, ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SCNN1G
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.09 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene SCNN1G
Gene Accession No. P37091, P51170, Q9WU39
Gene Alias [g]ENaC; amiloride sensitive sodium channel gamma1 subunit; amiloride-sensitive epithelial sodium channel gamma subunit; amiloride-sensitive sodium channel gamma-subunit; amiloride-sensitive sodium channel subunit gamma; BESC3; ENaC; ENaC gamma; ENaC gamma subunit; ENaCG; ENaCgamma; epithelial Na(+) channel subunit gamma; epithelial sodium channel gamma subunit; Gamma ENaC; Gamma NaCH; gamma-ENaC; Gamma-NaCH; nonvoltage-gated sodium channel 1 subunit gamma; PHA 1; PHA1; SCNEG; SCNN 1G; Scnn1g; sodium channe epithelial 1 gamma subunit; sodium channel epithelial 1 gamma subunit; Sodium channel nonvoltage-gated 1 gamma (epithelial); sodium channel, non voltage gated 1 gamma subunit; sodium channel, nonvoltage-gated 1 gamma; sodium channel, nonvoltage-gated 1, gamma; Sodium channel, nonvoltage-gated 1, gamma (epithelial); sodium channel, non-voltage-gated 1, gamma subunit; sodium channel, nonvoltage-gated, type I, gamma
Gene Symbols SCNN1G
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 100-250 of human SCNN1G (NP_001030.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20278, 24768, 6340
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.