Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma-ENaC Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109307
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109307 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109307 Supplier Invitrogen™ Supplier No. PA5109307
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: A431, 293T, rat kidney tissue, mouse brain tissue.

Epithelial sodium channels are amiloride-sensitive members of the degenerin/epithelial sodium channel (Deg/ENaC) superfamily of ion channels. Members of this superfamily of ion channels share organizational similarity in that they all possess two short intracellular amino and carboxyl termini, two short membrane spanning segments, and a large extracellular loop with a conserved cysteine-rich region. There are three homologous isoforms of the ENaC (alpha, beta, and gamma) protein. ENaC in the kidney, lung, and colon plays an essential role in trans-epithelial sodium and fluid balance. ENaC also mediates aldosterone-dependent sodium reabsorption in the distal nephron of the kidney, thus regulating blood pressure. ENaC is thought to be regulated, in part, through association with the cystic fibrosis transmembrane conductance regulator (CFTR) chloride ion channel. Gain-of-function mutations in beta- or gamma-ENaC can cause severe arterial hypertension (Liddel's syndrome) and loss-of-function mutations in alpha- or beta-ENaC causes pseudohypoaldosteronism (PHA-1).
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma-ENaC
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene SCNN1G
Gene Accession No. P51170
Gene Alias [g]ENaC; amiloride sensitive sodium channel gamma1 subunit; amiloride-sensitive epithelial sodium channel gamma subunit; amiloride-sensitive sodium channel gamma-subunit; amiloride-sensitive sodium channel subunit gamma; BESC3; ENaC; ENaC gamma; ENaC gamma subunit; ENaCG; ENaCgamma; epithelial Na(+) channel subunit gamma; epithelial sodium channel gamma subunit; Gamma ENaC; Gamma NaCH; gamma-ENaC; Gamma-NaCH; nonvoltage-gated sodium channel 1 subunit gamma; PHA 1; PHA1; SCNEG; SCNN 1G; Scnn1g; sodium channe epithelial 1 gamma subunit; sodium channel epithelial 1 gamma subunit; Sodium channel nonvoltage-gated 1 gamma (epithelial); sodium channel, non voltage gated 1 gamma subunit; sodium channel, nonvoltage-gated 1 gamma; sodium channel, nonvoltage-gated 1, gamma; Sodium channel, nonvoltage-gated 1, gamma (epithelial); sodium channel, non-voltage-gated 1, gamma subunit; sodium channel, nonvoltage-gated, type I, gamma
Gene Symbols SCNN1G
Host Species Rabbit
Immunogen Recombinant protein within Human SCNN1G aa 77-541.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6340
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles, store in dark
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.