Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma-Glutamylcyclotransferase/CRF21/GGCT Antibody (04), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP306551AF350
Description
gamma-Glutamylcyclotransferase/CRF21/GGCT Monoclonal antibody specifically detects gamma-Glutamylcyclotransferase/CRF21/GGCT in Human samples. It is validated for ELISASpecifications
gamma-Glutamylcyclotransferase/CRF21/GGCT | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
04 | |
ELISA | |
gamma-glutamylcyclotransferase, GCTG, GGC, MGC3077 | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human gamma-Glutamylcyclotransferase/CRF21/GGCT (O75223-1; Met1-Leu188). | |
0.1 mL | |
Apoptosis, Cell Biology, Endocrinology | |
79017 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction