Learn More
Keystone Cleanroom Products Gamma-Irradiated Bic Pen
SDP
Catalog No. p-7227830
Click to view available options
Color:
Black
Blue
This item is not returnable. View return policy
This item is not returnable. View return policy
Ready-to-use gamma irradiated pens and markers eliminate the risks associated with surface sanitization procedures.
  • Gamma irradiated
  • Lot controlled
  • Ready to use
  • Packaged in easy to open Tyvek™ peel pouches
  • Minimize contamination risks
  • Simplifies conformance to SOP's
  • Eliminate the need for wiping down contaminated writing instruments
Specifications
Product Type Cleanroom Pen
Color Black
Autoclavable Not Autoclavable
Sterility Sterile
Material Plastic
Disposable Single-use
Certifications/Compliance COC, Certificate of Irradiation
No. per Case 500/Cs.
Packaging Type Peel Pouch
Length (Metric) 14 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.