Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma Sarcoglycan Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19029925UL
Description
gamma Sarcoglycan Polyclonal specifically detects gamma Sarcoglycan in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
gamma Sarcoglycan | |
Polyclonal | |
Western Blot 0.4 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:20-1:50 | |
Q13326 | |
SGCG | |
This antibody was developed against Recombinant Protein corresponding to amino acids:YAKEIHSRVDSSLLLQSTQNVTVNARNSEGEVTGRLKVGPKMVEVQNQQFQINSNDGKPLFTVDEKEVVVGTDKLRVTGPEGALFEHSVETPLVRADPFQD | |
Affinity Purified | |
RUO | |
6445 | |
Human | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
35 kDa dystrophin-associated glycoprotein, 35DAG, A4, DAGA4,35kD dystrophin-associated glycoprotein, DMDA, DMDA1, gamma sarcoglycan, gamma-sarcoglycan, Gamma-SG, MAM, MGC130048, sarcoglycan, gamma (35kD dystrophin-associated glycoprotein), sarcoglycan, gamma (35kDa dystrophin-associated glycoprotein), SCARMD2, SCG3LGMD2C, TYPE | |
Rabbit | |
33 kDa | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction