Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
gamma-Synuclein Antibody (SAIC-31A-10), Alexa Fluor™ 350 , Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320099AF350
Description
gamma-Synuclein Monoclonal antibody specifically detects gamma-Synuclein in Human samples. It is validated for Western Blot, ELISASpecifications
gamma-Synuclein | |
Monoclonal | |
Alexa Fluor 350 | |
BCSG1PERSYN, Breast cancer-specific gene 1 protein, gamma-synuclein, persyn, SRPRSN, synoretin, synuclein, gamma (breast cancer-specific protein 1) | |
The KLH-conjugated peptide "e;YLAPSGPSGTLK"e;, derived from Gamma-synuclein. Gamma-synulein plays a role in neurofilament network integrity. | |
0.1 mL | |
Alzheimers Research, Cancer, Cellular Markers, Core ESC Like Genes, Mitotic Regulators, Neuroscience, Stem Cell Markers | |
6623 | |
Store at 4°C in the dark. | |
IgG κ |
Western Blot, ELISA | |
SAIC-31A-10 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction