Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Synuclein Monoclonal Antibody (1H10D2)
GREENER_CHOICE

Catalog No. PIMA515296
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515296 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515296 Supplier Invitrogen™ Supplier No. MA515296
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15296 targets SNCG (breast cancer-specific protein 1) in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15296 immunogen is purified recombinant fragment of SNCG expressed in E. Coli.

This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Synuclein
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1H10D2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene SNCG
Gene Accession No. O76070
Gene Alias BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1)
Gene Symbols SNCG
Host Species Mouse
Immunogen Purified recombinant fragment of SNCG expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.