Learn More
Invitrogen™ gamma Synuclein Recombinant Rabbit Monoclonal Antibody (4A0F4)

Description
Immunogen sequence: VVQSVTSVAE KTKEQANAVS EAVVSSVNTV ATKTVEEAEN IAVTSGVVRK EDLRPSAPQQ EGEASKEKEE VAEEAQSGGD.
Specifications
Specifications
| Antigen | gamma Synuclein |
| Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 4A0F4 |
| Concentration | 0.32 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SNCG |
| Gene Accession No. | O76070, Q63544, Q9Z0F7 |
| Gene Alias | BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1) |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.