Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ gamma Synuclein Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA519746
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA519746 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA519746 Supplier Invitrogen™ Supplier No. PA519746
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-19746 targets gamma Synuclein in WB applications and shows reactivity with Human samples. The PA5-19746 immunogen is synthetic peptide conjugated to KLH derived from within residues 100 to the C-terminus of Human gamma Synuclein. PA5-19746 detects gamma Synuclein which has a predicted molecular weight of approximately 15 kDa.

This gene encodes a member of the synuclein family of proteins which are believed to be involved in the pathogenesis of neurodegenerative diseases. Mutations in this gene have also been associated with breast tumor development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen gamma Synuclein
Applications Western Blot
Classification Polyclonal
Concentration 0.9 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.02% sodium azide; pH 7.4
Gene SNCG
Gene Accession No. O76070
Gene Alias BCSG1; Breast cancer-specific gene 1 protein; C79089; Gamma-synuclein; persyn; PRSN; Sensory neuron synuclein; SNCG; SR; Synoretin; synuclein gamma; synuclein, gamma; synuclein, gamma (breast cancer-specific protein 1)
Gene Symbols SNCG
Host Species Rabbit
Immunogen Synthetic peptide conjugated to KLH derived from within residues 100 to the C-terminus of Human gamma Synuclein.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6623
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.